missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 193 D7
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
193 D7 is a histone demethylase JMJD1C inhibitor (IC50 = 0.59 μM in in vitro demethylation assay). In vivo, 193 D7 suppresses tumor by targeting intratumoral Treg cells in mice tumor models. 193 D7 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 8-Benzoyl-2,3,6,7-tetrahydro-5-oxo-5H-thiazolo[3,2-a]pyridine-6-acetic acid |
| CAS | 861224-48-8 |
| Quantity | 50 mg |
| Target | Histone Demethylase Inhibitors |
| Molecular Formula | C16H15NO4S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction