Learn More
Agilent BioTek 192 Tube Wash Manifold for 384-well Microplates
GREENER_CHOICE
Product Code. 17834667 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
406 FX Washer Dispenser
Unit Size:
Each
Product Code. 17834667
Brand: Agilent BioTek 7180018
192 Tube Wash Manifold for use with the 406 FX Washer Dispenser.
  • Washes 384-well plates two times faster than standard 96 tube manifold included with 406 FX Washer Dispenser
TRUSTED_SUSTAINABILITY
Specifications
Type Manifold
For Use With (Equipment) 406 FX Washer Dispenser