Learn More
Fisherbrand™ 18mm PP Screw Seal, Black, open top, 18-400 thread, Assembled septum
Product Code. 12900961 Shop All Fisherbrand Products
Click to view available options
Color:
Blue Transparent, White, Black
Gray, Red, Black
Red, White, Black
Material (Liner):
PTFE/Butyl
Silicone/PTFE
Thickness (Metric):
1.5 mm
1.6 mm
1.7 mm
2 mm
Unit Size:
Pack of 100
Pack of 1000
Product Code. 12900961
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Pre-assembled caps and septa are convenient and minimize contamination from handling.
  • 18mm
  • Combination Seal
  • PP
  • Screw Cap
  • Black
  • Center hole
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE/Butyl
Color Gray, Red, Black
Diameter (Metric) Outer 18 mm
Material Polypropylene
For Use With (Application) Headspace
Quantity 100
Thickness (Metric) 2 mm
Type Screw Cap