Learn More
Equip Labo™ 180cm Headboard Wall
Product Code. 12552897
Change view
Click to view available options
Size: 180cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12552897 Size: 180cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12552897 Supplier Equip Labo™ Supplier No. 2069
  • ONLINE EXCLUSIVE PRICE
    52.70€
    44.08€ / Pack of 1
    Save 8.62€
    16% Off
 Restricted Item
Estimated Shipment: 20-11-2026