Learn More
Bochem™ 18/10 Stainless Steel Weighing Scoop
Product Code. p-8003336
Click to view available options
Length (Metric):
105 mm
105 mm, 235 mm
120 mm
200 mm, 330 mm
200 mm, 70 mm
250 mm, 120 mm
90 mm
Unit Size:
Each
Product Code. 10575221
Brand: Bochem™ 12812
Only null left
Specifications
Bottom Shape Conical
Material Stainless Steel
Shape Conical
Type Weighing Scoop
Length (Metric) 120 mm
Diameter (Metric) 32 mm