Learn More
Bochem™ 18/10 Stainless Steel Weighing Scoop
Brand:  Bochem™ 3602
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10231771
  • 15.75€ / Each
Estimated Shipment: 01-07-2024
Specifications
Conical
Conical
235 x 31 mm
31 mm
Stainless Steel
Weighing Scoop
105 mm, 235 mm
5 mm