Learn More
RSG Rostfrei Schneidwerkzeuge™ Stainless Steel Curved Spatulas
Click to view available options
Length (Metric):
150 mm
Unit Size:
Pack of 5
Description
- With plastic handles
- Available in a variety of end styles
Specifications
Specifications
| Quantity | 5 Pk. |
| Shape | Simple |
| Product Type | Curved Spatula |
| Handle Color | Black |
| Width (Metric) | 4 mm |
| Handle Material | Plastic |
| Material | Stainless Steel (18/10) |
| Handle Type | Plastic |
| Length (Metric) | 150 mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.