Learn More
Bochem™ 18/10 Stainless Steel Rods
Product Code. 10412331
Click to view available options
Length (Metric):
500 mm
600 mm
750 mm
1250 mm
1500 mm
For Use With (Equipment):
Stand bases
Stand bases with thread M10
Unit Size:
Each
Product Code. 10412331
Brand: Bochem™ 5118
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Material Stainless Steel (18/10)
Length (Metric) 1500 mm
Product Type Rod
For Use With (Application) For Stand Bases M10 Threaded
For Use With (Equipment) Stand bases with thread M10
Quantity 1 Ea.