Learn More
Bochem™ 18/10 Stainless Steel Propeller Stirrer
Product Code. 10060781
Click to view available options
Diameter (Metric):
70 mm
100 mm
Unit Size:
Each
Product Code. 10060781
Brand: Bochem™ 9058
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Description Propeller stirrer, 4 vertical blades
Diameter (Metric) 100 mm
Diameter (Metric) Shaft 10 mm
For Use With (Application) Mixing of media with low and middle viscosity
Length (Metric) 500 mm