Learn More
Bochem™ 18/10 Stainless Steel One-Sided Chemical Spoons
Click to view available options
Length (Metric):
120 mm
150 mm
180 mm
210 mm
300 mm
Unit Size:
Each
Description
- One-sided chemical spoon
- 18/10 stainless steel
Specifications
Specifications
| Quantity | 1 Ea. |
| Product Type | Chemical Scoop |
| Diameter (Metric) Outer | 5mm |
| Handle Material | Stainless Steel (18/10) |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 150 mm |