Learn More
Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas, Spoon Shape
Product Code. 10094950
Click to view available options
Length (Metric):
130 mm
180 mm
185 mm
210 mm
Product Type:
Micro Spoon Spatula
Spoon Spatula
Diameter (Metric) Outer:
2mm
2.5mm
3mm
4mm
Unit Size:
Each
Pack of 10
Product Code. 10094950
Brand: Bochem™ 3341
Edge
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas comes in two types.
  • 18/10 stainless steel
Specifications
Quantity 1 Ea.
Autoclavable Autoclavable
Shape Spoon
Product Type Micro Spoon Spatula
Length (Metric) Blade 40 mm
Width (Metric) 5 mm
Blade Material Stainless Steel (18/10)
Diameter (Metric) Outer 2.5mm
Dimensions (L x W) 180 x 5 mm
Material Stainless Steel (18/10)
Length (Metric) 180 mm