Learn More
Bochem™ 18/10 Stainless Steel Laboratory Spoon
Click to view available options
Length (Metric):
150 mm
170 mm
200 mm
Dimensions (L x W):
20 x 10 mm
30 x 15 mm
30 x 26 mm
40 x 10 mm
Unit Size:
Each
Description
- Lab spoon
- Manufactured from 18/10 stainless steel
Specifications
Specifications
| Quantity | 1 Ea. |
| Product Type | Laboratory Spoon |
| Dimensions (L x W) | 40 x 10 mm |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 170 mm |