Learn More
Bochem™ 18/10 Stainless Steel Laboratory Collapsible Trolley
Product Code. 12691905
Change view
Click to view available options
Dimensions (L x W x H):
785 x 450 x 850 mm (with wheel and handle)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
12691905 785 x 450 x 850 mm (with wheel and handle) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12691905 Supplier Bochem™ Supplier No. 11417
  • ONLINE EXCLUSIVE PRICE
    1725.00€
    1390.34€ / Pack of 1
    Save 334.66€
    19% Off
Estimated Shipment: 24-02-2027
Specifications
Dimensions (L x W x H) 785 x 450 x 850 mm (with wheel and handle)
Finish Matte
Height (Metric) 850 mm
Length (Metric) 785 mm
Load Bearing Capacity (Metric) 300 kg
Material Stainless Steel (18/10)
Width (Metric) 450 mm