Learn More
Agencinox™ 18/10 Stainless Steel High Evaporating Dish
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
5 L
Unit Size:
Each
Description
- High form evaporating dish
- Stainless steel construction
- Available in a variety of sizes
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 250 mL |
| Material | Stainless Steel |
| Shape | Round |
| Type | Evaporating Dish |
| For Use With (Application) | Evaporating Application |
| Height (Metric) | 45 mm |
| Diameter (Metric) | 85 mm |