Learn More
Bochem™ 18/10 Stainless Steel Evaporating Dish with Rim
Product Code. 10745505
Change view
Click to view available options
Height (Metric):
45 mm
60 mm
95 mm
100 mm
Length (Metric):
310 mm
340 mm
375 mm
445 mm
500 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Length (Metric) Unit Size
10745505 95 mm 445 mm Each
10346104 100 mm 500 mm Each
10098310 45 mm 310 mm Each
10162411 60 mm 340 mm Each
10739121 45 mm 375 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10745505 Supplier Bochem™ Supplier No. 8398
Estimated Shipment: 28-12-2026
Evaporate materials in the laboratory wit the Bochem™ 18/10 Stainless Steel Evaporating Dish with Rim.
  • 18/10 stainless steel
  • Substantial rim size
  • Rectangular shape
Specifications
Bottom Shape Flat
Material Stainless Steel
Shape Rectangular
Type Evaporating Dish
Length (Metric) 445 mm
Height (Metric) 95 mm