Learn More
Bochem™ 18/10 Stainless Steel Double Spatulas, Straight
Click to view available options
Length (Metric):
130 mm
150 mm
185 mm
210 mm
250 mm
300 mm
Unit Size:
Each
Description
- Double spatula
- Manufactured from 18/10 stainless steel
Specifications
Specifications
| Quantity | 1 Ea. |
| Shape | Straight |
| Coating | PTFE |
| Product Type | Double Ended Spatula |
| Length (Metric) Blade | 40 mm |
| Width (Metric) | 9 mm |
| Width (Metric) Blade | 9 mm |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 130 mm |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur