Learn More
Bochem™ 18/10 Stainless Steel Contact Bottom Evaporating Dishes
Product Code. 10271111
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Unit Size:
Each
Product Code. 10271111
Brand: Bochem™ 8563
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Bottom Shape Flat
Capacity (Metric) 1000 mL
Material Stainless Steel
Shape Round
Type Evaporating Dish
Height (Metric) 75 mm
Diameter (Metric) 140 mm