Learn More
Bochem™ 18/10 E-Poli Crucible Tongs With Ridges
Click to view available options
Length (Metric):
160 mm
200 mm
220 mm
250 mm
300 mm
400 mm
500 mm
600 mm
Unit Size:
Each
Description
- With ridges
- Stainless steel
Specifications
Specifications
| Length (Metric) | 400 mm |
| Material | Stainless Steel (18/10) |
| Type | Crucible Tongs |