Learn More
Bochem™ 18/10 E-Poli Crucible Tongs With Ridges
Product Code. 10140001
Change view
Click to view available options
Length (Metric):
160 mm
200 mm
220 mm
250 mm
300 mm
400 mm
500 mm
600 mm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10140001 300 mm Each
10788941 200 mm Each
10068800 160 mm Each
10232851 220 mm Each
10038840 250 mm Each
10282941 400 mm Each
10243041 500 mm Each
10554261 600 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10140001 Supplier Bochem™ Supplier No. 2103
  • ONLINE EXCLUSIVE PRICE
    12.75€
    10.33€ / Pack of 1
    Save 2.42€
    18% Off
Estimated Shipment: 30-11-2026
With ridges. Bochem™ 18/10 E-Poli Crucible Tongs With Ridges provided secure gripping.
  • With ridges
  • Stainless steel
Specifications
Product Type Crucible Tongs
Length (Metric) 300 mm
Material Stainless Steel (18/10)