Learn More
Bochem™ 18/10 E-Poli Crucible Tongs With Ridges
Product Code. 10068800
Click to view available options
Length (Metric):
160 mm
200 mm
220 mm
250 mm
300 mm
400 mm
500 mm
600 mm
Unit Size:
Each
Product Code. 10068800
Brand: Bochem™ 2099
With ridges. Bochem™ 18/10 E-Poli Crucible Tongs With Ridges provided secure gripping.
  • With ridges
  • Stainless steel
Specifications
Length (Metric) 160 mm
Material Stainless Steel (18/10)
Type Crucible Tongs