missing translation for 'onlineSavingsMsg'
Learn More

17Beta-Estradiol-13C2, TRC

Product Code. 18022000
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18022000 1 mg 1mg
18012200 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18022000 Supplier Toronto Research Chemicals Supplier No. TRCE8880031MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82938-05-4
Molecular Formula 13C2 C16 H24 O2
Molecular Weight (g/mol) 274.37
Synonym (17β)-Estra-1,3,5(10)-triene-3,17-diol-3,4-13C2, 17β-Estradiol-3,4-13C2
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES C[C@]12CC[C@H]3[C@@H](CCc4[13cH][13c](O)ccc34)[C@@H]1CC[C@@H]2O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1/i10+1,12+1
Formula Weight 274.1843
Percent Purity >95%
Chemical Name or Material 17beta-Estradiol-3,4-13C2
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.