Learn More
Cole Parmer Idex P-331 Super Flangeless Nut, Standard Knurl,Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom
Brand:  Cole Parmer 02021-80 
Idex P-331 Super Flangeless Nut, Standard Knurl,Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom
Product Code. 17083867
  • ONLINE EXCLUSIVE PRICE
    6.20€
    5.12€ / Each
    Save 1.08€
    17% Off
Only null left
Estimated Shipment: 17-02-2026