Learn More
Hamilton™ 1700 Series Standard PTFE Instrument Syringe
Product Code. p-8543006
Click to view available options
Volume (Metric):
12.5 μL
100 μL
250 μL
Unit Size:
Each
Product Code. 10490405
Brand: Hamilton™ 549522
Setting the standard for quality and reliability.
  • Syringe life is exceptional when used in conjunction with Hamilton™ precision syringe drives
  • Enables precision liquid handling
  • Gastight for leak-free performance
Specifications
Autoclavable Not Autoclavable
Diameter (Metric) Outer 7.75 mm
Max. Pressure 68.9 bar
Max. Temperature (Metric) 115°C
Model 1725.1
Quantity 1/Pk.
Volume (Metric) 100 μL
Connection Type Luer Lock