Learn More
17-beta-Hydroxy Exemestane, TRC
Product Code. 18080190
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18080190 5 mg 5mg
18082720 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18080190 Supplier Toronto Research Chemicals Supplier No. TRCH9423405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 122370-91-6
Molecular Formula C20 H26 O2
Molecular Weight (g/mol) 298.42
Synonym Androsta-1,4-dien-3-one, 17-hydroxy-6-methylene-, (17β)-, (17β)-17-Hydroxy-6-methyleneandrosta-1,4-dien-3-one, 17-Hydroexemestane, Hydroexemestane, 17β-Hydroxy Exemestane, 17β-Hydroxyexemestane
IUPAC Name (8R,9S,10R,13S,14S,17S)-17-hydroxy-10,13-dimethyl-6-methylidene-8,9,11,12,14,15,16,17-octahydro-7H-cyclopenta[a]phenanthren-3-one
SMILES C[C@]12CC[C@H]3[C@@H](CC(=C)C4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C20H26O2/c1-12-10-14-15-4-5-18(22)20(15,3)9-7-16(14)19(2)8-6-13(21)11-17(12)19/h6,8,11,14-16,18,22H,1,4-5,7,9-10H2,2-3H3/t14-,15-,16-,18-,19+,20-/m0/s1
Formula Weight 298.1933
Percent Purity >95%
Chemical Name or Material 17beta-Hydroxy Exemestane
Grade HPLC