missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17-beta-Estradiol D3 (16,16,17-D3) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 17beta-Estradiol-16,16,17-D3 |
| CAS | 79037-37-9 |
| Solvent or Matrix | Acetonitrile |
| Molecular Formula | C18 2H3 H21 O2 |
| Quantity | 1 mL |
| Synonym | Estra-1,3,5(10)-triene-16,16,17-d3-3,17-diol, (17β)-, (17β)-Estra-1,3,5(10)-triene-16,16,17-d3-3,17-diol, 17β-Estradiol-16,16,17-d3, Estradiol-d3, 17β-Estradiol-16,16,17-D3 |
| InChI Formula | InChI=1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1/i7D2,17D |
| SMILES | [2H]C1([2H])C[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]2(C)[C@@]1([2H])O |
| IUPAC Name | (8R,9S,13S,14S,17S)-16,16,17-trideuterio-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 275.4 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction