Learn More
17-alpha-Methylestradiol, TRC
Specifications
Specifications
| Chemical Name or Material | 17Alpha-Methylestradiol |
| CAS | 302-76-1 |
| Molecular Formula | C19 H26 O2 |
| Quantity | 50 mg |
| Synonym | 17-alpha-Methylestradiol, Estra-1,3,5(10)-triene-3,17-diol','17-methyl-','(17β)-, Estra-1,3,5(10)-triene-3,17β-diol','17-methyl- (7CI,8CI), Estradiol','17-methyl- (6CI), (17β)-17-Methylestra-1,3,5(10)-triene-3,17-diol, 17-Methyl-1,3,5(10)-estratriene-3,17β-diol, 17-Methylestradiol, 17 |
| InChI Formula | InChI=1S/C19H26O2/c1-18-9-7-15-14-6-4-13(20)11-12(14)3-5-16(15)17(18)8-10-19(18,2)21/h4,6,11,15-17,20-21H,3,5,7-10H2,1-2H3/t15-,16-,17+,18+,19+/m1/s1 |
| SMILES | C[C@]1(O)CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C |
| IUPAC Name | (8R,9S,13S,14S,17S)-13,17-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 286.409 |
| Formula Weight | 286.193 |