missing translation for 'onlineSavingsMsg'
Learn More

16alpha,17,21-Trihydroxy-pregna-1,4-diene-3,11,20-trione, TRC

Product Code. 30453527
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453527 Supplier Toronto Research Chemicals Supplier No. TRCT79523050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 16alpha,17,21-Trihydroxy-pregna-1,4-diene-3,11,20-trione
CAS 3754-05-0
Molecular Formula C21H26O6
Quantity 50 mg
Synonym Pregna-1,4-diene-3,11,20-trione','16,17,21-trihydroxy-','(16α)- (ACI), (16α)-16,17,21-Trihydroxypregna-1,4-diene-3,11,20-trione (ACI), Pregna-1,4-diene-3,11,20-trione','16α,17,21-trihydroxy- (6CI','7CI','8CI), 16a,17,21-Trihydroxy-pregna-1,4-diene-3,11,20-trione, 16α-Hydroxy-11-keto Prednisolone, (8S,9S,10R,13S,14S,16R,17S)-16,17-Dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,10,12,13,14,15,16,17-decahydro-3H-cyclopenta[a]phenanthrene-3,11(6H)-dione), 16α,17,21-Trihydroxypregna-1,4-diene-3,11,20-trione
InChI Formula InChI=1S/C21H26O6/c1-19-6-5-12(23)7-11(19)3-4-13-14-8-16(25)21(27,17(26)10-22)20(14,2)9-15(24)18(13)19/h5-7,13-14,16,18,22,25,27H,3-4,8-10H2,1-2H3/t13-,14-,16+,18+,19-,20-,21-/m0/s1
SMILES C[C@]12CC(=O)[C@H]3[C@H]([C@@H]1C[C@H]([C@@]2(C(=O)CO)O)O)CCC4=CC(=O)C=C[C@]34C
IUPAC Name (8S,9S,10R,13S,14S,16R,17S)-16,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione
Molecular Weight (g/mol) 374.43
Formula Weight 374.1729
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.