Learn More
Corning™ 1536-Well Nontreated Polystyrene Plates
Product Code. 10465823 Shop All Corning Products
Change view
Click to view available options
Color:
Black
White
Volume (Metric) Working:
8 μL
10 μL
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) Working Unit Size
10465823 White 8 μL Pack of 50
10795683 Black 10 μL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10465823 Supplier Corning™ Supplier No. 3725
  • ONLINE EXCLUSIVE PRICE
    1245.00€
    1160.14€ / Pack of 50
    Save 84.86€
    6% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
Bulk Quote Request
Superior flatness minimizes variability within a plate and between plates
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer™ FlexDrop™
  • Working volume of up to 10 μL
Generic Bar Code
  • Ensures compatibility with most bar code scanning and software systems
  • To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface)
  • Labels applied to all four sides of the plate No sequence number duplicates
Order Info
Packaged in bags of 10
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 32 x 48
Color White
For Use With (Equipment) Automated dispensers
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Flat Bottom (F-well)
For Use With (Application) For barcode scanning and software systems
No. per Case 50/Cs.
No. per Pack 10/Pk.