Learn More
Corning™ 1536-Well Black/Clear Round Bottom Ultra-low Attachment Spheroid Microplate with Lid, Sterile
Shop All Corning ProductsDescription
- 1536-well format allows for high throughput 3D cell culture
- Optically clear round bottom with black opaque microplate body
- Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
- Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
- Unique design shields each well to minimize well-to-well cross-talk.
- You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate.
- Compatible with the GNF ultra high throughput system
Specifications
Specifications
| Lid | With Lid |
| Color | Black, Clear Bottom |
| Volume (Metric) Well | 14 mL |
| No. of Wells | 1536 |
| Volume (Metric) Working | 5 to 10 μL |
| Sterility | Sterile |
| Surface Treatment | Ultra-low Binding |
| Well Shape | Round Bottom (U-well) |
| Well Color | Clear |