Learn More
Corning™ 1536-Well Black/Clear Bottom Not Treated Low Base Polystyrene Microplate without Lid, Non-sterile
Shop All Corning ProductsDescription
- Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
- Recommended working volume up to 8 μL
- Round well bottom for reduced air entrapment and improved CV values and Z factor
- Raised well bottom for higher sensitivity
- Flood reservoir on four sides to reduce instrument contamination
- Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
Specifications
Specifications
| Lid | Without Lid |
| Color | Black, Clear Bottom |
| Material | Polystyrene |
| No. of Wells | 1536 |
| Volume (Metric) Working | Up to 8 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Quantity | 10/Pk., 50/Cs. |