Learn More
14-Methyl-7,8,13b,14-tetrahydroindolo[2',3':3,4]pyrido[2,1-b]quinazolin-5(13H)-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 518-18-3 |
| Molecular Formula | C19H17N3O |
| MDL Number | MFCD00016673 |
| Quantity | 1 g |
| SMILES | O=C1N2C(C(NC3=C4C=CC=C3)=C4CC2)N(C)C5=C1C=CC=C5 |
| Molecular Weight (g/mol) | 303.36 |
| Percent Purity | 98% |
| Chemical Name or Material | 14-Methyl-7,8,13b,14-tetrahydroindolo[2',3':3,4]pyrido[2,1-b]quinazolin-5(13H)-one |