Learn More
Welch Ilmvac™ 14 Laborzentrifugen Oil
Product Code. 10652963 Shop All Welch Ilmvac Products
Change view
Click to view available options
Volume (Metric):
2 L
Unit Size:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10652963 2 L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10652963 Supplier Welch Ilmvac™ Supplier No. 800136
  • ONLINE EXCLUSIVE PRICE
    113.00€
    111.80€ / 2 L
    Save 1.20€
    1% Off
Estimated Shipment: 16-10-2026