Learn More
Fisherbrand™ 13mm Crimp Seal, Silver, Centre Tear-Off, assembled septum
Product Code. p-7113944 Shop All Fisherbrand Products
Click to view available options
Color:
Blue, Gray
Gray, Red
Unit Size:
Pack of 1000
Product Code. 11874960
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Aluminum crimp closures provide a secure leak-resistant seal.
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Aluminum seals must be applied with a crimping tool
TRUSTED_SUSTAINABILITY
Specifications
Description Pharma-Fix-Septa
Material (Liner) PTFE/Butyl
Color Blue, Gray
Diameter (Metric) Outer 13 mm
Material Aluminum
For Use With (Application) HPLC
Quantity 100
Thickness (Metric) 2 mm
Type Crimp Cap