Learn More
13(S)-HODE, TRC
Specifications
Specifications
| Chemical Name or Material | 13(S)-HODE |
| CAS | 29623-28-7 |
| Molecular Formula | C18H32O3 |
| Quantity | 5 mg |
| Synonym | 13(S)-Hydroxyoctadeca-9Z,11E-dienoic Acid, (9Z,11E,13S)-13-Hydroxy-9,11-octadecadienoic Acid, (+)-Coriolic Acid, (13S,9Z,11E)-13-Hydroxy-9,11-octadecadienoic Acid, (9Z,11E,13S)-13-Hydroxyoctadecadienoic Acid, (S)-Coriolic Acid, 13S-HODE, 13S-Hydroxy-9Z,11E-octadecadienoic Acid, L-13-Hydroxy-cis-9,trans-11-octadecadienoic Acid |
| InChI Formula | InChI=1S/C18H32O3/c1-2-3-11-14-17(19)15-12-9-7-5-4-6-8-10-13-16-18(20)21/h7,9,12,15,17,19H,2-6,8,10-11,13-14,16H2,1H3,(H,20,21)/b9-7-,15-12+/t17-/m0/s1 |
| SMILES | OC(CCCCCCC/C=C\C=C\[C@@H](O)CCCCC)=O |
| IUPAC Name | (S,9Z,11E)-13-hydroxyoctadeca-9,11-dienoic acid |
| Molecular Weight (g/mol) | 296.44 |
| Formula Weight | 296.24 |