Learn More
11,12-Didehydro Retinol, TRC
Specifications
Specifications
| Chemical Name or Material | 11,12-Didehydro Retinol |
| CAS | 29443-88-7 |
| Molecular Formula | C20 H28 O |
| Quantity | 10 mg |
| Synonym | 3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,6,8-nonatrien-4-yn-1-ol, 11,12-Didehydroretinol |
| InChI Formula | InChI=1S/C20H28O/c1-16(8-6-9-17(2)13-15-21)11-12-19-18(3)10-7-14-20(19,4)5/h8,11-13,21H,7,10,14-15H2,1-5H3/b12-11+,16-8+,17-13+ |
| SMILES | C\C(=C/C#C\C(=C\CO)\C)\C=C\C1=C(C)CCCC1(C)C |
| IUPAC Name | (2E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,6,8-trien-4-yn-1-ol |
| Molecular Weight (g/mol) | 284.436 |
| Formula Weight | 284.214 |