Learn More
Medchem Express 10mM * 1 mL PK14105
Brand:  Medchem Express HY-100346_1mL
Encompass_Preferred
10mM * 1 mL PK14105
Product Code. 18611483
  • 137.70€ / 1mL
 Restricted Item
Estimated Shipment: 25-11-2026