Learn More
Macherey Nagel 10ML DMF-DMA, 1X10 ML
Brand:  Macherey Nagel 701430.110
10ML DMF-DMA, 1X10 ML
Product Code. 16572510
  • ONLINE EXCLUSIVE PRICE
    71.75€
    64.70€ / 10mL
    Save 7.05€
    9% Off
Currently Unavailable: We have no information when this product will be available.
View alternative products
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • DMF-DMA, 1x10 mL
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Supplemental information
  • MIXTURE LIST-Contains : N,N-dimethylformamide dimethylacetal