missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vonoprazan Fumarate-D3, TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | Vonoprazan Fumarate-D3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17H13D3FN3O2S • C4H4O4 |
| Quantity | 10 mg |
| Synonym | TAK 438-D3, Takecab-D3, 5-(2-Fluorophenyl)-N-methyl-1-(3-pyridinylsulfonyl)-1H-pyrrole-3-methanamine 2-Butenedioate-D3 |
| InChI Formula | InChI=1S/C17H16FN3O2S.C4H4O4/c1-19-10-13-9-17(15-6-2-3-7-16(15)18)21(12-13)24(22,23)14-5-4-8-20-11-14;5-3(6)1-2-4(7)8/h2-9,11-12,19H,10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/i1D3; |
| SMILES | FC1=CC=CC=C1C2=CC(CNC([2H])([2H])[2H])=CN2S(C3=CC=CN=C3)(=O)=O.OC(/C=C/C(O)=O)=O |
| IUPAC Name | N-((5-(2-fluorophenyl)-1-(pyridin-3-ylsulfonyl)-1H-pyrrol-3-yl)methyl)methan-d3-amine fumarate |
| Molecular Weight (g/mol) | 464.48 |
| Formula Weight | 464.12 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction