missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Capsaicin-d3 (E/Z-Mixture), TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Capsaicin O-methyl-d3 (E,Z) |
| CAS | 1185237-43-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 D3 H24 N O3 |
| Quantity | 10 mg |
| Synonym | N-[(4-Hydroxy-3-(methoxy-d3)phenyl)methyl]-8-methyl-6-nonenamide, (E/Z)-8-Methyl-N-vanillyl-6-nonenamide-d3 |
| InChI Formula | InChI=1S/C18H27NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h6,8,10-12,14,20H,4-5,7,9,13H2,1-3H3,(H,19,21)/i3D3 |
| SMILES | [2H]C([2H])([2H])Oc1cc(CNC(=O)CCCCC=CC(C)C)ccc1O |
| IUPAC Name | N-[[4-hydroxy-3-(trideuteriomethoxy)phenyl]methyl]-8-methyl-non-6-enamide |
| Molecular Weight (g/mol) | 308.43 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction