Learn More
Baclofen-d4, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Baclofen phenyl-d4 |
| CAS | 1189938-30-4 |
| Molecular Formula | C10 2H4 H8 Cl N O2 |
| Quantity | 10 mg |
| Synonym | β-(Aminomethyl)-4-chloro-benzenepropanoic Acid-d4, β-(Aminomethyl)-p-chlorohydrocinnamic Acid-d4, β-(4-Chlorophenyl)-GABA-d5, Ba-34647-d4, Baclon-d4, Clofen-d4, Lioresal-d4 |
| InChI Formula | InChI=1S/C10H12ClNO2/c11-9-3-1-7(2-4-9)8(6-12)5-10(13)14/h1-4,8H,5-6,12H2,(H,13,14)/i1D,2D,3D,4D |
| SMILES | [2H]c1c([2H])c(C(CN)CC(=O)O)c([2H])c([2H])c1Cl |
| IUPAC Name | 4-amino-3-(4-chloro-2,3,5,6-tetradeuteriophenyl)butanoic acid |
| Molecular Weight (g/mol) | 217.69 |
| Formula Weight | 217.0808 |