Learn More
Azilsartan-d5, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Azilsartan-d5 |
| CAS | 1346599-45-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C25 H15 D5 N4 O5 |
| Quantity | 10 mg |
| Synonym | Azilsartan-D5 (ethyl-D5), Azilsartan-d5, 3-[[4-[2-(5-Oxo-4H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]-2-(1,1,2,2,2-pentadeuterioethoxy)benzimidazole-4-carboxylic acid, 3-[[4-[2-(5-Oxo-4H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]-2-(ethoxy-1,1,2,2,2-d5)benzimidazole-4-carboxylic acid |
| InChI Formula | InChI=1S/C25H20N4O5/c1-2-33-24-26-20-9-5-8-19(23(30)31)21(20)29(24)14-15-10-12-16(13-11-15)17-6-3-4-7-18(17)22-27-25(32)34-28-22/h3-13H,2,14H2,1H3,(H,30,31)(H,27,28,32)/i1D3,2D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])Oc1nc2cccc(C(=O)O)c2n1Cc3ccc(cc3)c4ccccc4C5=NOC(=O)N5 |
| IUPAC Name | 3-[[4-[2-(5-oxo-4H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]-2-(1,1,2,2,2-pentadeuterioethoxy)benzimidazole-4-carboxylic acid |
| Molecular Weight (g/mol) | 461.482 |
| Show More |