missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Dimethylamino)ethyl]-1-indanone (>90%), TRC

Product Code. 30003290
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003290 10 mg 10mg
30133268 50 mg 50mg
30132652 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003290 Supplier Toronto Research Chemicals Supplier No. TRCD46128010MG
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3409-21-0
Molecular Formula C13 H17 N O
Molecular Weight (g/mol) 203.2802
Synonym 2-[2-(Dimethylamino)ethyl]-2,3-dihydro-1H-inden-1-one
IUPAC Name 2-[2-(dimethylamino)ethyl]-2,3-dihydroinden-1-one
SMILES CN(C)CCC1Cc2ccccc2C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H17NO/c1-14(2)8-7-11-9-10-5-3-4-6-12(10)13(11)15/h3-6,11H,7-9H2,1-2H3
Formula Weight 203.131
Chemical Name or Material 2-[2-(Dimethylamino)ethyl]-1-indanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.