missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Aminopropyl)-n-dodecylpropane-1,3-diamine (>80%), TRC

Product Code. 30342943
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30342943

Brand: Toronto Research Chemicals TRCA89475010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3-Aminopropyl)-n-dodecylpropane-1,3-diamine
CAS 2372-82-9
Purity Grade Notes GC
Molecular Formula C18 H41 N3
Quantity 10 g
Synonym 1,3-Propanediamine, N-(3-aminopropyl)-N-dodecyl- (9CI), Dodecylamine, N,N-bis(3-aminopropyl)- (6CI,7CI,8CI), N1-(3-Aminopropyl)-N1-dodecyl-1,3-propanediamine, Bis(3-aminopropyl)dodecylamine, Grotan BA 21, Lonzabac 12, Lonzabac 12.100, Lonzabac 12.30, Lonzabac 1230, Mistral, N,N-Bis(3-aminopropyl)dodecylamine, N,N-Bis(3-aminopropyl)laurylamine, RC 5637, Triameen Y 12, Triameen Y 12D, Triamine Y 12D
InChI Formula InChI=1S/C18H41N3/c1-2-3-4-5-6-7-8-9-10-11-16-21(17-12-14-19)18-13-15-20/h2-20H2,1H3
SMILES CCCCCCCCCCCCN(CCCN)CCCN
IUPAC Name N'-(3-aminopropyl)-N'-dodecylpropane-1,3-diamine
Molecular Weight (g/mol) 299.54
Formula Weight 299.33
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.