Learn More
L-Valine Amide, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | L-Valine Amide |
| CAS | 3014-80-0 |
| Molecular Formula | C5H13ClN2O |
| Quantity | 10 g |
| Synonym | 2(S)- 2-Amino-3-methylbutanamide Monohydrochloride, (S)-2-Amino-3-methylbutanamide Monohydrochloride,, L-Monohydrochloride Valinamide, (2S)-2-Amino-3-methylbutanamide Hydrochloride, (S)-2-Amino-3-methylbutanamide Hydrochloride, (S)-Valinamide Hydrochloride, L-Valinamide Hydrochloride, L-Valine Amide Hydrochloride |
| InChI Formula | InChI=1S/C5H12N2O.ClH/c1-3(2)4(6)5(7)8;/h3-4H,6H2,1-2H3,(H2,7,8);1H/t4-;/m0./s1 |
| SMILES | Cl.CC(C)[C@H](N)C(=O)N |
| IUPAC Name | (2S)-2-amino-3-methylbutanamide;hydrochloride |
| Molecular Weight (g/mol) | 152.62 |
| Formula Weight | 152.0716 |