missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol-(OD)3, CDN
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Specifications
Specifications
| Chemical Name or Material | Glycerol-O,O,O-D3 |
| CAS | 7325-16-8 |
| Molecular Formula | C3 D3 H5 O3 |
| Quantity | 10 g |
| Synonym | 1,2,3-Propanetriol-1,2,3-d3 (ACI), 1,2,3-Propanetriol-d3 (9 CI), Glycerol-O,O,O-d3 (8 CI), 1,2,3-Trideuteriooxypropane, Glycerine-d3, Glycerol-d3, Glycerol-(OD)3, Glycerol-O,O,O-D3 |
| InChI Formula | InChI=1 S/C3H8O3/c4-1-3(6)2-5/h3-6 H,1-2H2/i4D,5 D,6 D |
| SMILES | [2 H]OCC(CO[2 H])O[2 H] |
| IUPAC Name | 1,2,3-trideuteriooxypropane |
| Molecular Weight (g/mol) | 95.1123 |
| Formula Weight | 95.0662 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction