Learn More
Ammonium-d4 Chloride, CDN
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Ammonium Chloride-d4 |
| CAS | 12015-14-4 |
| Molecular Formula | Cl . D4 N |
| Quantity | 10 g |
| Synonym | Ammonium chloride ((ND4)Cl) (7 CI,8 CI,9 CI), Ammonium chloride-d4, Ammonium-d4 chloride, Deuterated ammonium chloride (ND4Cl), Hydrochloric acid-d, mono(ammoniate-d3), Tetradeuteroammonium chloride, Ammonium chloride-d4 |
| InChI Formula | InChI=1 S/ClHH3N/h1H;1H3/i/hD4 |
| SMILES | [Cl-].[2 H][N+]([2 H])([2 H])[2 H] |
| IUPAC Name | tetradeuterioazanium;chloride |
| Molecular Weight (g/mol) | 57.516 |
| Formula Weight | 57.028 g/mol |
| Show More |