Learn More
Chem-Lab 100ML Rhodium standard solution (Plasma HIQU)(2.558 g RhCl3.3H2O / l 10% HCl)
Brand: Chem-Lab CL01.1811.0100
100ML Rhodium standard solution (Plasma HIQU)(2.558 g RhCl3.3H2O / l 10% HCl)
Safety and Handling
- Rhodium standard solution 10 µg/ml
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.