missing translation for 'onlineSavingsMsg'
Learn More

Zoalene, TRC

Product Code. 17922935
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922935 100 mg 100mg
17982935 25 mg 25mg
17933635 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17922935 Supplier Toronto Research Chemicals Supplier No. TRCZ584000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148-01-6
Molecular Formula C8 H7 N3 O5
Molecular Weight (g/mol) 225.16
Synonym Benzamide, 2-methyl-3,5-dinitro-, o-Toluamide, 3,5-dinitro- (6CI,8CI), 2-Methyl-3,5-dinitrobenzamide, 3,5-Dinitro-2-methylbenzamide, 3,5-Dinitro-o-toluamide, Coccidin, Coccidine A, Coccidot, Dinitolmide, Salcostat, Zoalene, Zoamix
IUPAC Name 2-methyl-3,5-dinitrobenzamide
SMILES Cc1c(cc(cc1[N+](=O)[O-])[N+](=O)[O-])C(=O)N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H7N3O5/c1-4-6(8(9)12)2-5(10(13)14)3-7(4)11(15)16/h2-3H,1H3,(H2,9,12)
Formula Weight 225.0386
Percent Purity >95%
Chemical Name or Material 3,5-Dinitro-o-toluamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.