Learn More
Veratric Acid 4-Chlorobutyl Ester, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Veratric Acid 4-Chlorobutyl Ester |
| CAS | 69788-75-6 |
| Molecular Formula | C13H17ClO4 |
| Quantity | 100 mg |
| Synonym | 3,4-Dimethoxybenzoic Acid 4-Chlorobutyl Ester |
| InChI Formula | InChI=1S/C13H17ClO4/c1-16-11-6-5-10(9-12(11)17-2)13(15)18-8-4-3-7-14/h5-6,9H,3-4,7-8H2,1-2H3 |
| SMILES | COc1ccc(cc1OC)C(=O)OCCCCCl |
| IUPAC Name | 4-chlorobutyl 3,4-dimethoxybenzoate |
| Molecular Weight (g/mol) | 272.72 |
| Formula Weight | 272.0815 |