Learn More
Vaccenic Acid, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | trans-Vaccenic Acid |
| CAS | 693-72-1 |
| Molecular Formula | C18 H34 O2 |
| Quantity | 100 mg |
| Synonym | 11-Octadecenoic acid','(E)- (8CI), Vaccenic acid (6CI), (11E)-11-Octadecenoic acid, (E)-11-Octadecenoic acid, (E)-Octadec-11-enoic acid, 11(E)-Octadecenoic acid, 11-trans-Octadecenoic acid, trans-11-Octadecenoic acid, trans-Vaccenic acid, trans-Δ11-Octadecenoic acid, trans-11-Octadecanoic Acid, 11-Octadecenoic acid','(11E)- |
| InChI Formula | InChI=1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h7-8H,2-6,9-17H2,1H3,(H,19,20)/b8-7+ |
| SMILES | CCCCCC\C=C\CCCCCCCCCC(=O)O |
| IUPAC Name | (E)-octadec-11-enoic acid |
| Molecular Weight (g/mol) | 282.46 |
| Formula Weight | 282.2559 |